4-Hexylresorcinol, 98%
4-Hexylresorcinol, 98%
4-Hexylresorcinol, 98%
4-Hexylresorcinol, 98%
Thermo Scientific Chemicals
4-Hexylresorcinol, 98%
CAS: 136-77-6 | C12H18O2 | 194.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA197920250
Price (MYR)
422.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
422.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Hexylresorcinol
Name Note99%
CAS136-77-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning