Fluoroboric acid, 50 wt% solution in water
Fluoroboric acid, 50 wt% solution in water
Fluoroboric acid, 50 wt% solution in water
Fluoroboric acid, 50 wt% solution in water
Thermo Scientific Chemicals
Fluoroboric acid, 50 wt% solution in water
CAS: 16872-11-0 | BF4H | 87.812 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA196705000
Price (MYR)
688.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
688.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFluoroboric acid
ColorWhite
CAS16872-11-0
Melting Point-90.0°C
Boiling Point130.0°C
View more
C-X bond formation reagent
RUO – Research Use Only