Iodine, 99.5%, extra pure, resublimed
Iodine, 99.5%, extra pure, resublimed
Iodine, 99.5%, extra pure, resublimed
Iodine, 99.5%, extra pure, resublimed
Thermo Scientific Chemicals
Iodine, 99.5%, extra pure, resublimed
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
View all
Glass bottle
Catalog number FSA196565000
Price (MYR)
1,250.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,250.00
EA
Add to cart
Specifications
Chemical Name or MaterialIodine
CAS7553-56-2
ColorGray
Health Hazard 1Warning
Health Hazard 2GHS P Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
View more
C-X bond formation reagent
RUO – Research Use Only