Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
Thermo Scientific Chemicals
Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA196435000
Price (MYR)
1,179.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,179.00
BOT
Add to cart
Chemical Identifiers
CAS7440-62-2, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialVanadium standard solution
Name Note1 mg/ml V in 2% HNO3
PackagingPlastic bottle
CAS7440-62-2
Health Hazard 1GHS Signal Word: Warning
View more
for atomic adsorption spectroscopy
RUO – Research Use Only