Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3
Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3
Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3
Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3
Thermo Scientific Chemicals
Nickel standard solution, for AAS, 1 mg/ml Ni in 2-5% HNO3
CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA196155000
Price (MYR)
575.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
575.00
BOT
Add to cart
Chemical Identifiers
CAS7440-02-0, 13478-00-7, 7697-37-2, 7732-18-5
Specifications
Chemical Name or MaterialNickel standard solution
Name Note1 mg/ml Ni in 2% HNO3
PackagingPlastic bottle
CAS7440-02-0
Health Hazard 1Danger
View more
For atomic adsorption spectroscopy
RUO – Research Use Only