Gold standard solution, for AAS, 1 mg/ml Au in 10-20% HCl
Gold standard solution, for AAS, 1 mg/ml Au in 10-20% HCl
Gold standard solution, for AAS, 1 mg/ml Au in 10-20% HCl
Gold standard solution, for AAS, 1 mg/ml Au in 10-20% HCl
Gold standard solution, for AAS, 1 mg/ml Au in 10-20% HCl
CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA196005000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5, 7647-01-0, 7732-18-5
Specifications
Chemical Name or MaterialGold standard solution
Name Note1 mg/ml Au in 0.5N HCl
PackagingPlastic bottle
CAS7440-57-5
Health Hazard 1Danger
View more
For atomic adsorption spectroscopy
RUO – Research Use Only