Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Thermo Scientific Chemicals
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
CAS: 10141-05-6 | CoN2O6 | 182.941 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA195925000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS10141-05-6, 7732-18-5
Specifications
Chemical Name or MaterialCobalt standard solution
Name Note1 mg/ml Co in 2% HNO3
PackagingPlastic bottle
CAS10141-05-6
Health Hazard 1Danger
View more
For atomic adsorption spectroscopy
RUO – Research Use Only