Boron standard solution, for AAS, 1 mg/ml B in water
Boron standard solution, for AAS, 1 mg/ml B in water
Boron standard solution, for AAS, 1 mg/ml B in water
Boron standard solution, for AAS, 1 mg/ml B in water
Thermo Scientific Chemicals
Boron standard solution, for AAS, 1 mg/ml B in water
CAS: 7440-42-8 | B | 10.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA195865000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS7440-42-8
Specifications
Chemical Name or MaterialBoron standard solution
PackagingPlastic bottle
CAS7440-42-8
RTECS NumberED7350000
EINECS Number231-151-2
View more
For atomic adsorption spectroscopy
RUO – Research Use Only