2-Acetyl-1-naphthol, 99%
2-Acetyl-1-naphthol, 99%
2-Acetyl-1-naphthol, 99%
2-Acetyl-1-naphthol, 99%
Thermo Scientific Chemicals
2-Acetyl-1-naphthol, 99%
CAS: 711-79-5 | C12H10O2 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA195000250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Acetyl-1-naphthol
Name Note99%
CAS711-79-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning