Ammonium iodide, ACS reagent
Ammonium iodide, ACS reagent
Ammonium iodide, ACS reagent
Ammonium iodide, ACS reagent
Thermo Scientific Chemicals
Ammonium iodide, ACS reagent
CAS: 12027-06-4 | H4IN | 144.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA193212500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium iodide
Melting Point551.0°C
CAS12027-06-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement