Ruthenium Red tetrahydrate, pure
Ruthenium Red tetrahydrate, pure
Ruthenium Red tetrahydrate, pure
Ruthenium Red tetrahydrate, pure
Thermo Scientific Chemicals
Ruthenium Red tetrahydrate, pure
CAS: 12790-48-6 | Cl6H36N14O6Ru3-14 | 844.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA191730010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialRuthenium red tetrahydrate
Name NotePure
CAS12790-48-6
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.