Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Thermo Scientific Chemicals
Fluorescein diacetate, 97%, pure
CAS: 596-09-8 | C24H16O7 | 416.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA191660250
Price (MYR)
1,272.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,272.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFluorescein diacetate
CAS596-09-8
Melting Point200.0°C to 205.0°C
ColorYellow
EINECS Number209-877-6