2-Bromo-6-methoxynaphthalene, 98%
2-Bromo-6-methoxynaphthalene, 98%
2-Bromo-6-methoxynaphthalene, 98%
2-Bromo-6-methoxynaphthalene, 98%
Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%
CAS: 5111-65-9 | C11H9BrO | 237.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA190560250
Price (MYR)
762.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
762.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Bromo-6-methoxynaphthalene
Name Note98%
CAS5111-65-9
CAS Min %97.5
CAS Max %100.0