View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2,3,5,6-Tetrafluorophenol
Name Note98%
CAS769-39-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger