tert-Butylchlorodiphenylsilane, 98%
tert-Butylchlorodiphenylsilane, 98%
tert-Butylchlorodiphenylsilane, 98%
tert-Butylchlorodiphenylsilane, 98%
Thermo Scientific Chemicals
tert-Butylchlorodiphenylsilane, 98%
CAS: 58479-61-1 | C16H19ClSi | 274.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA187500500
Price (MYR)
1,020.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,020.00
BOT
Add to cart
Specifications
Chemical Name or Materialtert-Butylchlorodiphenylsilane
CAS58479-61-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at