1-Bromotetradecane, 98%
1-Bromotetradecane, 98%
1-Bromotetradecane, 98%
1-Bromotetradecane, 98%
Thermo Scientific Chemicals
1-Bromotetradecane, 98%
CAS: 112-71-0 | C14H29Br | 277.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA187385000
Price (MYR)
1,436.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,436.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Bromotetradecane
Name Note98%
CAS112-71-0
CAS Min %97.5
CAS Max %100.0