1,3-Dibromobenzene, 97%
1,3-Dibromobenzene, 97%
1,3-Dibromobenzene, 97%
1,3-Dibromobenzene, 97%
Thermo Scientific Chemicals
1,3-Dibromobenzene, 97%
CAS: 108-36-1 | C6H4Br2 | 235.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Packaging:
Glass bottle
Catalog number FSA186640500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 3-Dibromobenzene
Name Note97%
CAS108-36-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning