Tryptamine, 98%
Tryptamine, 98%
Tryptamine, 98%
Thermo Scientific Chemicals
Tryptamine, 98%
CAS: 61-54-1 | C10H12N2 | 160.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA186122500
Price (MYR)
2,736.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,736.00
EA
Add to cart
Specifications
Chemical Name or MaterialTryptamine
CAS61-54-1
Melting Point113.0°C to 117.0°C
ColorWhite
EINECS Number200-510-5