1,4-Diiodobenzene, 98%
1,4-Diiodobenzene, 98%
1,4-Diiodobenzene, 98%
1,4-Diiodobenzene, 98%
Thermo Scientific Chemicals
1,4-Diiodobenzene, 98%
CAS: 624-38-4 | C6H4I2 | 329.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA185970500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 4-Diiodobenzene
CAS624-38-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0