Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™
Thermo Scientific Chemicals
Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™
CAS: 563-43-9 | C2H5AlCl2 | 126.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA185451000
Price (MYR)
775.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
775.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthylaluminium dichloride
CAS563-43-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effec
CAS142-82-5
View more
Lewis acid
RUO – Research Use Only