Pyridinium chlorochromate, 98%
Pyridinium chlorochromate, 98%
Pyridinium chlorochromate, 98%
Pyridinium chlorochromate, 98%
Thermo Scientific Chemicals
Pyridinium chlorochromate, 98%
CAS: 26299-14-9 | C5H6ClCrNO3 | 215.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA183675000
Price (MYR)
883.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
883.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPyridinium chlorochromate
Name Note98%
CAS26299-14-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause cancer by inhalation.
May intensify fire; oxidizer.