Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™
Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™
Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™
Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™
Thermo Scientific Chemicals
Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™
CAS: 75-16-1 | CH3BrMg | 119.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA183541000
Price (MYR)
396.00
ML
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
396.00
ML
Add to cart
Specifications
Chemical Name or MaterialMethylmagnesium bromide
ColorColorless to Yellow
Name Note3M Solution in Diethyl Ether
CAS75-16-1
Health Hazard 1Danger