2,3-Dihydroxybenzaldehyde, 97%
2,3-Dihydroxybenzaldehyde, 97%
2,3-Dihydroxybenzaldehyde, 97%
2,3-Dihydroxybenzaldehyde, 97%
Thermo Scientific Chemicals
2,3-Dihydroxybenzaldehyde, 97%
CAS: 24677-78-9 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA183510250
Price (MYR)
1,071.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,071.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 3-Dihydroxybenzaldehyde
Name Note97%
CAS24677-78-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning