D(+)-Trehalose dihydrate, 99%
D(+)-Trehalose dihydrate, 99%
D(+)-Trehalose dihydrate, 99%
D(+)-Trehalose dihydrate, 99%
Thermo Scientific Chemicals
D(+)-Trehalose dihydrate, 99%
CAS: 6138-23-4 | C12H26O13 | 378.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA182555000
Price (MYR)
5,017.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,017.00
EA
Add to cart
Specifications
Chemical Name or MaterialD(+)-Trehalose dihydrate
CAS6138-23-4
Health Hazard 1Warning
Health Hazard 2May form combustible dust concentrations in air
Melting Point97.0°C to 99.0°C