Furan, 99+%, stabilized
Furan, 99+%, stabilized
Furan, 99+%, stabilized
Furan, 99+%, stabilized
Thermo Scientific Chemicals
Furan, 99+%, stabilized
CAS: 110-00-9 | C4H4O | 68.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1811100252.5 LGlass bottle
Catalog number FSA181110025
Price (MYR)
953.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
953.00
EA
Add to cart
Specifications
Chemical Name or MaterialFuran
CAS110-00-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Harmful if inhaled.
May cause cancer.
Harmful to aquatic life with long
CAS128-37-0