Propionitrile, 99%, extra pure
Propionitrile, 99%, extra pure
Propionitrile, 99%, extra pure
Propionitrile, 99%, extra pure
Thermo Scientific Chemicals
Propionitrile, 99%, extra pure
CAS: 107-12-0 | C3H5N | 55.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA180895000
Price (MYR)
704.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
704.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPropionitrile
CAS107-12-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled.
Fatal in contact with skin.
Fatal if swallowed.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
View more