1,1,2,2-Tetrabromoethane, 98%
1,1,2,2-Tetrabromoethane, 98%
1,1,2,2-Tetrabromoethane, 98%
1,1,2,2-Tetrabromoethane, 98%
Thermo Scientific Chemicals
1,1,2,2-Tetrabromoethane, 98%
CAS: 79-27-6 | C2H2Br4 | 345.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number FSA180872500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1,1,2,2-Tetrabromoethane
CAS79-27-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0