Alginic acid, sodium salt
Alginic acid, sodium salt
Alginic acid, sodium salt
Alginic acid, sodium salt
Thermo Scientific Chemicals
Alginic acid, sodium salt
CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
250 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA177775000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAlginic acid, sodium salt
CAS9005-38-3
RTECS NumberAZ5820000
TSCATSCA
Molecular Formula(C6H7O7)A(C6H7O7)BNa