2-Naphthoic acid, 99%
2-Naphthoic acid, 99%
2-Naphthoic acid, 99%
2-Naphthoic acid, 99%
Thermo Scientific Chemicals
2-Naphthoic acid, 99%
CAS: 93-09-4 | C11H8O2 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA177321000
Price (MYR)
1,308.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,308.00
EA
Add to cart
Specifications
Chemical Name or Material2-Naphthoic acid
CAS93-09-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0