Undecanoic acid, 99%
Undecanoic acid, 99%
Undecanoic acid, 99%
Undecanoic acid, 99%
Thermo Scientific Chemicals
Undecanoic acid, 99%
CAS: 112-37-8 | C11H22O2 | 186.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA173971000
Price (MYR)
958.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
958.00
BOT
Add to cart
Specifications
Chemical Name or MaterialUndecanoic acid
Name Note99%
CAS112-37-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: