Pentadecafluorooctanoic acid, 96%
Pentadecafluorooctanoic acid, 96%
Pentadecafluorooctanoic acid, 96%
Pentadecafluorooctanoic acid, 96%
Pentadecafluorooctanoic acid, 96%
CAS: 335-67-1 | C8HF15O2 | 414.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA173961000
Price (MYR)
0.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
0.00
EA
Add to cart
Specifications
Chemical Name or MaterialPentadecafluorooctanoic acid
CAS335-67-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Suspected of causing cancer.
May damage the unborn child.
May cause harm to breast-fed children.
Causes damage to organs throu
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre