5-Nitro-2-furaldehyde, 97%
5-Nitro-2-furaldehyde, 97%
5-Nitro-2-furaldehyde, 97%
5-Nitro-2-furaldehyde, 97%
Thermo Scientific Chemicals
5-Nitro-2-furaldehyde, 97%
CAS: 698-63-5 | C5H3NO4 | 141.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA173860100
Price (MYR)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material5-Nitro-2-furaldehyde
CAS698-63-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0