Phenyl disulfide, 99%
Phenyl disulfide, 99%
Phenyl disulfide, 99%
Phenyl disulfide, 99%
Thermo Scientific Chemicals
Phenyl disulfide, 99%
CAS: 882-33-7 | C12H10S2 | 218.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number FSA173712500
Price (MYR)
1,245.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,245.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenyl disulfide
Name Note99%
CAS882-33-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.