3-Methylsalicylic acid, 99%
3-Methylsalicylic acid, 99%
3-Methylsalicylic acid, 99%
3-Methylsalicylic acid, 99%
Thermo Scientific Chemicals
3-Methylsalicylic acid, 99%
CAS: 83-40-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA173341000
Price (MYR)
229.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
229.00
EA
Add to cart
Specifications
Chemical Name or Material3-Methylsalicylic acid
CAS83-40-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0