Sodium methoxide, pure, anhydrous powder
Sodium methoxide, pure, anhydrous powder
Sodium methoxide, pure, anhydrous powder
Sodium methoxide, pure, anhydrous powder
Thermo Scientific Chemicals
Sodium methoxide, pure, anhydrous powder
CAS: 124-41-4 | CH3NaO | 54.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA173125000
Price (MYR)
660.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
660.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium methoxide
Alkalinitytotal alkalinity (as NaOCH3):> 99%
CAS124-41-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Self-heating; may catch fire.
Flammable solid.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.