Heptafluorobutyric acid, 99%
Heptafluorobutyric acid, 99%
Heptafluorobutyric acid, 99%
Heptafluorobutyric acid, 99%
Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%
CAS: 375-22-4 | C4HF7O2 | 214.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA172805000
Price (MYR)
3,966.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,966.00
EA
Add to cart
Specifications
Chemical Name or MaterialHeptafluorobutyric acid
CAS375-22-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre