Bicine, 99+%, for analysis
Bicine, 99+%, for analysis
Bicine, 99+%, for analysis
Bicine, 99+%, for analysis
Thermo Scientific Chemicals
Bicine, 99+%, for analysis
CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Glass bottle
Catalog number FSA172652500
Price (MYR)
907.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
907.00
EA
Add to cart
Specifications
Absorbance0.05 max. (5% aqueous soln.) at 250nm
Chemical Name or MaterialBicine
Absorbance Conditions(5% aq. soln.) (1cm cell)
Name Note99+%
CAS150-25-4
View more
Biological buffer
RUO – Research Use Only