MOPS, 99%, for biochemistry
MOPS, 99%, for biochemistry
MOPS, 99%, for biochemistry
MOPS, 99%, for biochemistry
Thermo Scientific Chemicals
MOPS, 99%, for biochemistry
CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA172635000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Absorbance0.03 max. at 280nm
Chemical Name or MaterialMOPS
Absorbance Conditions(1M aq. soln.) (1 cm cell)
Name Note99%
CAS1132-61-2
View more
Biological buffer
RUO – Research Use Only