PIPES, 98.5+%, for biochemistry
PIPES, 98.5+%, for biochemistry
PIPES, 98.5+%, for biochemistry
PIPES, 98.5+%, for biochemistry
Thermo Scientific Chemicals
PIPES, 98.5+%, for biochemistry
CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA172611000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Absorbance(0.1M in 0.1M NaOH) (1 cm cell), (0.1M in 0.1M NaOH) (1 cm cell) at 0nm, 0.04 max. at 280nm, 0.05 max. at 260nm
Chemical Name or MaterialPIPES
Absorbance Conditions(0.1M in 0.1M NaOH) (1cm cell)
CAS5625-37-6
ColorWhite
View more
Biological buffer
RUO – Research Use Only