NBD chloride, 98%
NBD chloride, 98%
NBD chloride, 98%
Thermo Scientific Chemicals
NBD chloride, 98%
CAS: 10199-89-0 | C6H2ClN3O3 | 199.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA172390050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNBD chloride
Melting Point96.0°C to 99.0°C
CAS10199-89-0
CAS Min %97.5
CAS Max %100.0