3-Aminopyrazole, 98%
3-Aminopyrazole, 98%
3-Aminopyrazole, 98%
3-Aminopyrazole, 98%
Thermo Scientific Chemicals
3-Aminopyrazole, 98%
CAS: 1820-80-0 | C3H5N3 | 83.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Packaging:
Glass bottle
Catalog number FSA171030100
Price (MYR)
1,297.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,297.00
EA
Add to cart
Specifications
Chemical Name or Material3-Aminopyrazole
CAS1820-80-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0