Ethidium bromide, 95%, pure
Ethidium bromide, 95%, pure
Ethidium bromide, 95%, pure
Ethidium bromide, 95%, pure
Thermo Scientific Chemicals
Ethidium bromide, 95%, pure
CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA170960250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthidium bromide
CAS1239-45-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing genetic defects.
Fatal if inhaled.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.