9-Fluorenylmethyl chloroformate, 98%
9-Fluorenylmethyl chloroformate, 98%
9-Fluorenylmethyl chloroformate, 98%
9-Fluorenylmethyl chloroformate, 98%
Thermo Scientific Chemicals
9-Fluorenylmethyl chloroformate, 98%
CAS: 28920-43-6 | C15H11ClO2 | 258.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA170941000
Price (MYR)
5,150.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,150.00
EA
Add to cart
Specifications
Chemical Name or Material9-Fluorenylmethyl chloroformate
CAS28920-43-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Coupling reagent
RUO – Research Use Only