Paraformaldehyde, 90%, pure
Paraformaldehyde, 90%, pure
Paraformaldehyde, 90%, pure
Paraformaldehyde, 90%, pure
Thermo Scientific Chemicals
Paraformaldehyde, 90%, pure
CAS: 30525-89-4 | CH2O | 30.026 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA169650010
Price (MYR)
298.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
298.00
BOT
Add to cart
Specifications
Chemical Name or MaterialParaformaldehyde
CAS30525-89-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
Harmful if inhaled.<br
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF