4-Dimethylaminobenzaldehyde, 99+%
4-Dimethylaminobenzaldehyde, 99+%
4-Dimethylaminobenzaldehyde, 99+%
4-Dimethylaminobenzaldehyde, 99+%
Thermo Scientific Chemicals
4-Dimethylaminobenzaldehyde, 99+%
CAS: 100-10-7 | C9H11NO | 149.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA168765000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Dimethylaminobenzaldehyde
CAS100-10-7
Health Hazard 1GHS Signal Word: Warning
Melting Point70°C to 75°C
ColorGreen to Yellow