Cyclobutanol, 99+%
Cyclobutanol, 99+%
Cyclobutanol, 99+%
Thermo Scientific Chemicals
Cyclobutanol, 99+%
CAS: 2919-23-5 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
5 g
Packaging:
Glass bottle
Catalog number FSA168752500
Price (MYR)
304.00
BOT
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
304.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCyclobutanol
CAS2919-23-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.