3-Piperidinemethanol, 98%
3-Piperidinemethanol, 98%
3-Piperidinemethanol, 98%
3-Piperidinemethanol, 98%
Thermo Scientific Chemicals
3-Piperidinemethanol, 98%
CAS: 4606-65-9 | C6H13NO | 115.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA168040050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Piperidinemethanol
CAS4606-65-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0