N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%
Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%
CAS: 25561-30-2 | C8H18F3NOSi2 | 257.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Glass Bottle
Catalog number FSA168001000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN, O-Bis(trimethylsilyl)trifluoroacetamide
CAS25561-30-2
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Silylation reagent
RUO – Research Use Only