Decanoic acid, 99%
Decanoic acid, 99%
Decanoic acid, 99%
Thermo Scientific Chemicals
Decanoic acid, 99%
CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA167275000
Price (MYR)
817.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
817.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDecanoic acid
Name Note99+%
CAS334-48-5
Melting Point30.0°C to 32.0°C
Boiling Point268.0°C to 270.0°C