Adonitol, 98%
Adonitol, 98%
Adonitol, 98%
Thermo Scientific Chemicals
Adonitol, 98%
Adonitol, 98%, C5H12O5, CAS Number-488-81-3, adonit, adonite, adonitol, d-ribitol, d-xylitol, eutrit, ribitol, xylit, xylite, xylitol, 100g, 97.5% min. (ELSD) (HPLC), 01,53, 100.0, 99.0, White, 207-685-7, 152.15, HEBKCHPVOIAQTA-NGQZWQHPSA-N, Authentic, (2S,4R)-pentane-1,2,3,4,5-pentol | CAS: 488-81-3 | C5H12O5 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA167031000
Price (MYR)
1,888.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,888.00
EA
Add to cart
Specifications
Chemical Name or MaterialAdonitol
CAS488-81-3
CAS Min %99.0
CAS Max %100.0
Melting Point101.0°C to 104.0°C